Orders LinkContactUsLinkComponent
Beakers, low form
Beakers, low form
Supplier:   Lenz Laborglas
Beakers, low form
Supplier:  Lenz Laborglas
Certificates
Beakers, low form
Beakers, low form
14 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Graduated for approximate reading of contents and labelling area (exept 5 ml beaker, not graduated).
Certifications: DIN 12331 and ISO 3819
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 5 ml
    30 mm
    23 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 77,70 €
    77,70 €
    1 * 10 ST
       77,70 €
  • 25 ml
    50 mm
    34 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 75,00 €
    75,00 €
    1 * 10 ST
       75,00 €
  • 50 ml
    60 mm
    42 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 72,50 €
    72,50 €
    1 * 10 ST
       72,50 €
  • 100 ml
    70 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 72,10 €
    72,10 €
    1 * 10 ST
       72,10 €
  • 150 ml
    80 mm
    60 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 75,30 €
    75,30 €
    1 * 10 ST
       75,30 €
  • 250 ml
    95 mm
    70 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 78,10 €
    78,10 €
    1 * 10 ST
       78,10 €
  • 400 ml
    110 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 93,80 €
    93,80 €
    1 * 10 ST
       93,80 €
  • 600 ml
    125 mm
    90 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 117,00 €
    117,00 €
    1 * 10 ST
       117,00 €
  • 800 ml
    135 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 161,00 €
    161,00 €
    1 * 10 ST
       161,00 €
  • 1000 ml
    145 mm
    105 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 186,00 €
    186,00 €
    1 * 10 ST
       186,00 €
  • 2000 ml
    185 mm
    132 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 324,00 €
    324,00 €
    1 * 10 ST
       324,00 €
  • 3000 ml
    210 mm
    152 mm
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 235,00 €
    235,00 €
    1 * 4 ST
       235,00 €
  • 5000 ml
    270 mm
    170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 95,50 €
    95,50 €
    1 * 1 ST
       95,50 €
  • 10000 ml
    350 mm
    217 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 264,00 €
    264,00 €
    1 * 1 ST
       264,00 €