Pasteur pipettes
Supplier: HIRSCHMANN
Jump to:
About this item
Glass pasteur pipettes with fine drawn tips.
- With or without cotton plug
- Short or long form
Certifications: According to ISO 7712.
4 Options Available Below
Product Details & Documents
Glass pasteur pipettes with fine drawn tips.
- With or without cotton plug
- Short or long form
Certifications: According to ISO 7712.
Documents
HIRS9250201_TDS_EN HIRS9260101_TDS_EN
Datasheet
Product Family Options
Short form
Product Information
- CapacityLengthPackedAvailabilityPrice
- HIRS9250101Non-plugged, tip ∼45 mm longNon-plugged, tip ∼45 mm long2,0 ml150 mm250/sleeve, 4 sleeves/caseLog in for availability1 * 1.000 ST69,10 €
- HIRS9250201Plugged, tip ∼45 mm longPlugged, tip ∼45 mm long2,0 ml150 mm250/sleeve, 4 sleeves/caseLog in for availability1 * 1.000 ST104,00 €
Long form
Product Information
- CapacityLengthPackedAvailabilityPrice
- HIRS9260101Non-plugged, tip ∼120 mm longNon-plugged, tip ∼120 mm long2,0 ml230 mm250/sleeve, 4 sleeves/caseLog in for availability1 * 1.000 ST78,00 €
- HIRS9260201Plugged, tip ∼120 mm longPlugged, tip ∼120 mm long2,0 ml230 mm250/sleeve, 4 sleeves/caseLog in for availability1 * 1.000 ST113,00 €