Orders LinkContactUsLinkComponent
Burette stopcocks, straight
Burette stopcocks, straight
Supplier:   Brand
Burette stopcocks, straight
Supplier:  Brand
Certificates
Burette stopcocks, straight
Burette stopcocks, straight
2 Options Available Below
Product Details & Documents
Borosilicate glass 5,4, amber.
  • Interchangeable key
  • With glass or PTFE key
  • Key with retention device
Stopcock housing with burette tip and key.
Product Family Options
With PTFE key
Product Information
  • For
    Availability
    Price
  • Burettes of 25 ml capacity
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 55,50 €
    55,50 €
    1 * 1 ST
       55,50 €
  • Burettes of 50 ml capacity
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 55,50 €
    55,50 €
    1 * 1 ST
       55,50 €
Spare Keys
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Spare PTFE key for 25 ml and 50 ml burette capacity
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,80 €
    24,80 €
    1 * 1 ST
       24,80 €