Orders LinkContactUsLinkComponent
Burette stopcocks, lateral
Burette stopcocks, lateral
Supplier:   Brand
Burette stopcocks, lateral
Supplier:  Brand
Certificates
Burette stopcocks, lateral
Burette stopcocks, lateral
3 Options Available Below
Product Details & Documents
Borosilicate glass 5,4, amber.
  • Interchangeable key
  • With glass or PTFE key
  • Key with retention device
Stopcock housing with burette tip and key.
Product Family Options
Stopcocks
Product Information
  • For
    Availability
    Price
  • BRND81927
    Stopcock with PTFE key
    Stopcock with PTFE key
    Burettes of 25 and 50 ml capacity
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57,40 €
    57,40 €
    1 * 1 ST
       57,40 €
Spare Keys
Product Information
  • For
    Availability
    Price
  • BRND81167
    Spare PTFE key for 25 ml and 50 ml burette capacity
    Spare PTFE key for 25 ml and 50 ml burette capacity
    Automatic burette, Pellet pattern
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,70 €
    37,70 €
    1 * 1 ST
       37,70 €
  • BRND81248
    Spare PTFE key for 25 ml and 50 ml burette capacity
    Spare PTFE key for 25 ml and 50 ml burette capacity
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,80 €
    24,80 €
    1 * 1 ST
       24,80 €