AdvanTip™ Pipette Tips
Supplier: HAMILTON BONADUZ
Jump to:
About this item
AdvanTip™ precision pipette tips are perfect for routine, high-throughput work where an extensive range of universal tips and multiple packaging options are needed to meet laboratory requirements.
- Made from chemically resistant, virgin PP for excellent clarity and smooth surfaces
- Certified free from RNase, DNase, and pyrogens
- Extended length (only some tips): Prevent sample cross-contamination and protect pipettes from contamination when pipetting into long, narrow tubes or deep well plates
- Low retention tips (only some tips): Feature walls that maximise sample recovery; ideal for quantitative PCR, valuable samples and reagents
- Universal fit, can be used with Hamilton SoftGrip™ manual pipettes
4 Options Available Below
Product Details & Documents
AdvanTip™ precision pipette tips are perfect for routine, high-throughput work where an extensive range of universal tips and multiple packaging options are needed to meet laboratory requirements.
- Made from chemically resistant, virgin PP for excellent clarity and smooth surfaces
- Certified free from RNase, DNase, and pyrogens
- Extended length (only some tips): Prevent sample cross-contamination and protect pipettes from contamination when pipetting into long, narrow tubes or deep well plates
- Low retention tips (only some tips): Feature walls that maximise sample recovery; ideal for quantitative PCR, valuable samples and reagents
- Universal fit, can be used with Hamilton SoftGrip™ manual pipettes
Product Family Options
Product Information
- Tip styleVolumeSterileFor use withPackedAvailabilityPrice
- Standard2 - 300 µl-250, 300 μl pipettesRacked, 96 tips/rackLog in for availability1 * 960 ST95,20 €
- Low retention2 - 300 µl-250, 300 μl pipettesBulk, 1000 tips/bagLog in for availability1 * 1.000 ST99,50 €
- Low retention2 - 300 µl-250, 300 μl pipettesRacked, 96 tips/rackLog in for availability1 * 960 ST119,00 €
- Low retention2 - 300 µl+250, 300 μl pipettesRacked, 96 tips/rackLog in for availability1 * 960 ST134,00 €