Orders LinkContactUsLinkComponent
Microplates, 384-well
Supplier:   HIRSCHMANN
Microplates, 384-well
Supplier:  HIRSCHMANN
Certificates
Microplates, 384-well
3 Options Available Below
Product Details & Documents
Highly pure, chemically inert aluminium plate for individual glass inserts. Translucent plastic cover included.
Product Family Options
Product Information
  • Well volume
    Availability
    Price
  • 391-1971
    Microplate, PP, flat bottom, 384-well, with fixed glass inserts, chemically inert
    Microplate, PP, flat bottom, 384-well, with fixed glass inserts, chemically inert
    ~250 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 262,00 €
    262,00 €
    1 * 1 ST
       262,00 €
  • 612-9873
    Microplate, PP, flat bottom, deep shape, 384-well, with loose glass inserts, chemically inert
    Microplate, PP, flat bottom, deep shape, 384-well, with loose glass inserts, chemically inert
    ~250 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 262,00 €
    262,00 €
    1 * 1 ST
       262,00 €
  • HIRS9242184
    Microplate, aluminium, flat bottom, 384-well, with loose glass inserts, chemically inert
    Microplate, aluminium, flat bottom, 384-well, with loose glass inserts, chemically inert
    ~500 µl
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 267,00 €
    267,00 €
    1 * 1 ST
       267,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Glass inserts for 384 well plates
    Glass inserts for 384 well plates
    HIRS9240584
    Log in for availability
    1 * 384 ST - 118,00 €