Orders LinkContactUsLinkComponent
O-Ring seals for flat flanges
O-Ring seals for flat flanges
Supplier:   Lenz Laborglas
O-Ring seals for flat flanges
Supplier:  Lenz Laborglas
Certificates
O-Ring seals for flat flanges
O-Ring seals for flat flanges
8 Options Available Below
Product Details & Documents
Silicone or Silicone PTFE-coated.
  • Excellent chemical resistance
  • Excellent temperature resistance
Seals for flat flanges LF 60-200.
Product Family Options
Silicone
Product Information
  • For
    Availability
    Price
  • Flat flanges DN 60
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,60 €
    10,60 €
    1 * 1 ST
       10,60 €
  • Flat flanges DN 100
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,10 €
    17,10 €
    1 * 1 ST
       17,10 €
  • Flat flanges DN 150
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,70 €
    33,70 €
    1 * 1 ST
       33,70 €
  • Flat flanges DN 200
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,60 €
    41,60 €
    1 * 1 ST
       41,60 €
Silicone, PTFE-coated
Product Information
  • For
    Availability
    Price
  • Flat flanges DN 60
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,90 €
    31,90 €
    1 * 1 ST
       31,90 €
  • Flat flanges DN 100
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,40 €
    37,40 €
    1 * 1 ST
       37,40 €
  • Flat flanges DN 150
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,40 €
    45,40 €
    1 * 1 ST
       45,40 €
  • Flat flanges DN 200
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,60 €
    77,60 €
    1 * 1 ST
       77,60 €