ContactUsLinkComponent
Pipette stand, rotary
Pipette stand, rotary
Catalog # HIRS8902101
Supplier:  HIRSCHMANN
undefined
Pipette stand, rotary
Catalog # HIRS8902101
Supplier:  HIRSCHMANN
Specifications
  • Ø×H:
    470×230 mm
  • For:
    94 volumetric pipettes
  • Colour:
    Blue
  • Description:
    Pipette stand, rotary
  • Material:
    PP
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Pipette stand made from blue PP.
  • Rotatable model with a reinforced base and an additional, rotatable disk to protect the pipette tips
  • 94 holes with different diameters for graduated and volumetric pipettes of any size