Orders LinkContactUsLinkComponent
Stopcocks for desiccators
Stopcocks for desiccators
Needle-valve stopcock, PTFE, cone size 24/29, aperture 0-4 mm
Catalog # BRND82415
Supplier:   Brand
Stopcocks for desiccators
Catalog # BRND82415
Supplier:  Brand
Stopcocks for desiccators
Stopcocks for desiccators
Needle-valve stopcock, PTFE, cone size 24/29, aperture 0-4 mm
Price
53,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stopcocks made of borosilicate glass 3.3 are suitable for Novus desiccators with lid socket.
  • Lateral venting helps to reduce turbulence
  • Nozzle with outer Ø 10 mm
Specifications
  • Description:
    Needle-valve stopcock, PTFE, cone size 24/29, aperture 0-4 mm