Orders LinkContactUsLinkComponent
Paste, silicone
Paste, silicone
Silicone paste, low viscosity
Catalog # BRND61605
Supplier:   Brand
Paste, silicone
Catalog # BRND61605
Supplier:  Brand
Paste, silicone
Paste, silicone
Silicone paste, low viscosity
Price
39,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
For greasing ground joints.
  • Low viscosity
  • Operating temperature –40 to +160 °C
Specifications
  • Description:
    Silicone paste, low viscosity