Orders LinkContactUsLinkComponent
Stopcocks for burettes, straight
Stopcocks for burettes, straight
Supplier:   HIRSCHMANN
Stopcocks for burettes, straight
Supplier:  HIRSCHMANN
Certificates
Stopcocks for burettes, straight
Stopcocks for burettes, straight
3 Options Available Below
Product Details & Documents
Straight burette stopcock, made of borosilicate glass, with glass key for burettes, clear glass.
  • Excellent chemical resistance
  • Excellent temperature resistance
Product Family Options
Product Information
  • For
    Availability
    Price
  • 2 - 10 ml burettes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 55,20 €
    55,20 €
    1 * 1 ST
       55,20 €
  • 25 ml burettes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 55,20 €
    55,20 €
    1 * 1 ST
       55,20 €
  • 50 ml burettes
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 55,20 €
    55,20 €
    1 * 1 ST
       55,20 €