Orders LinkContactUsLinkComponent
Pycnometer heads with/without bottle
Pycnometer heads with/without bottle
Supplier:   Lenz Laborglas
Pycnometer heads with/without bottle
Supplier:  Lenz Laborglas
Certificates
Pycnometer heads with/without bottle
Pycnometer heads with/without bottle
4 Options Available Below
Product Details & Documents
Set, made of DURAN® borosilicate glass, consists of wide neck bottle with conical shoulder and NS joint, and pycnometer head.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Suitable for density determinations of bitumen-like and mineral materials.
Documents
Product Family Options
Bottle with head
Product Information
  • Volume (nom.)
    NS
    Availability
    Price
  • 500 ml
    45/27
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 98,00 €
    98,00 €
    1 * 1 ST
       98,00 €
  • 1000 ml
    60/37
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 111,00 €
    111,00 €
    1 * 1 ST
       111,00 €
Head only
Product Information
  • Volume (nom.)
    NS
    Availability
    Price
  • LENZ05660206
    Head for 500 ml bottle
    Head for 500 ml bottle
    45/27
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,10 €
    65,10 €
    1 * 1 ST
       65,10 €
  • LENZ05660207
    Head for 1 litre bottle
    Head for 1 litre bottle
    60/37
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 75,40 €
    75,40 €
    1 * 1 ST
       75,40 €
Head only
Product Information
  • Catalog #
    Description
    NS
    Availability
    Price
  • Head for 500 ml bottle
    45/27
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 65,10 €
    65,10 €
    1 * 1 ST
       65,10 €
Product Information
  • Catalog #
    Description
    NS
    Availability
    Price
  • Head for 1 litre bottle
    60/37
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 75,40 €
    75,40 €
    1 * 1 ST
       75,40 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Bottle for pycnometer NS60/46, 1000 ml
    Bottle for pycnometer NS60/46, 1000 ml
    LENZ07690070
    Log in for availability
    1 * 1 ST - 68,60 €