Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, 1-Nonene
Supelco® Organic Reference Standard, 1-Nonene
Catalog # 74323-5ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, 1-Nonene
Catalog # 74323-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    124-11-8
  • Description:
    1-Nonene
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H226 - H304 - H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00009568
  • M-Clarity quality level:
    MQ100
  • Melting point:
    –81 °C (lit.)
  • Formula:
    CH₃(CH₂)₆CH=CH₂
  • Molecular weight:
    126,24
  • Density:
    0,73 g/ml at 25 °C (lit.)
  • Flash point:
    24 °C - Closed cup
  • Boiling point:
    146 °C (lit.)

Specifications

About this item

  • GHS signal: GHS02, GHS08, GHS07
Caution: P210 - P301 + P310 + P331 - P302 + P352 - P305 + P351 + P338