Order Entry
Belgium
ContactUsLinkComponent
Tetrachloroethylene, Uvasol® for spectroscopy, Supelco®
Tetrachloroethylene, Uvasol® for spectroscopy, Supelco®
Supplier:  Merck
Certificates
undefined
Tetrachloroethylene, Uvasol® for spectroscopy, Supelco®
Supplier:  Merck
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Solubility:
    0.16 g/L (20 °C)
  • Melting Point:
    -22 °C
  • Molar Mass:
    165.83 g/mol
  • Boiling Point:
    121 °C (1013 hPa)
  • Vapor Pressure:
    19 hPa (20 °C)
  • Density:
    1.62 g/cm³ (20 °C)
  • pH:
    (H₂O) not applicable
  • Saturation concentration:
    126 g/m³ (20 °C) Air
  • Evaporation number:
    11
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

Specification Test Results

Related Information

Frequently Bought Together