Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sedimentation cones, Imhoff type

Sedimentation cones, Imhoff type

Imhoff Cones 1000 ml, with screw cap for emtying
Catalog # HIRS7720193
Supplier:   HIRSCHMANN
Sedimentation cones, Imhoff type
Catalog # HIRS7720193
Supplier:  HIRSCHMANN
Sedimentation cones, Imhoff type
Sedimentation cones, Imhoff type

Imhoff Cones 1000 ml, with screw cap for emtying

Price
151,00 €
Log in for availability
1 * 3 ST
Product Details & Documents

Sedimentation cone, Imhoff, made of SAN, clear and unbreakable with elevated scale, screw cap for rapid emptying.

  • As transparent as glass
  • Easy draining via screw plug

Graduation: 0 to 2 ml:0,1/ 2 to 10 ml:0,5/ 10 to 40 ml:1/ 40 to 100 ml:2/ 100 to 1000 ml:10

Specifications
  • Type:
    with screw cap for emtying
  • Description:
    Imhoff Cones 1000 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.