Orders LinkContactUsLinkComponent
Drying tubes, U-shaped
Drying tubes, U-shaped
Supplier:   Lenz Laborglas
Drying tubes, U-shaped
Supplier:  Lenz Laborglas
Certificates
Drying tubes, U-shaped
Drying tubes, U-shaped
3 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Drying tubes, U-shaped, side tubes with olives (Ext.Ø: 6 mm).
Documents
Product Family Options
Product Information
  • Length
    Ø int.
    Availability
    Price
  • 100 mm
    13 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,20 €
    27,20 €
    1 * 1 ST
       27,20 €
  • 130
    13 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,20 €
    27,20 €
    1 * 1 ST
       27,20 €
  • 180
    18 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,50 €
    29,50 €
    1 * 1 ST
       29,50 €