ContactUsLinkComponent
Measuring beakers with handle
Measuring beakers with handle
Catalog # HIRS7600286
Supplier:  HIRSCHMANN
Supplier #: 7600286
undefined
Measuring beakers with handle
Catalog # HIRS7600286
Supplier:  HIRSCHMANN
Supplier #: 7600286
Log in for availability
Select an Option
70 mm
Select an Option
5 ml
Select an Option
250 ml
Select an Option
120 mm
Select an Option
03
Select an Option
12
About this item
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to +121 °C (for 20 minutes), prolonged use +100 °C
  • Smooth surfaces for easy, fast cleaning
  • With spout
Ideal for general laboratory use and a wide variety of other applications. With embossed scale, blue graduations.
Food-safe products according to EC Directive No. 10/2011.
Frequently Bought Together