Orders LinkContactUsLinkComponent
Beakers, low form
Supplier:   HIRSCHMANN
Beakers, low form
Supplier:  HIRSCHMANN
Certificates
Beakers, low form
Beakers, low form
13 Options Available Below
Product Details & Documents
PP, highly translucent.
  • Very good chemical resistance
  • Cleaning up to maximum +60 °C is recommended to preserve marks and inscriptions
  • For autoclaving (+121 °C) beakers with an embossed scale should be selected
With printed blue scale, beaded rim and spout.
 : ISO 7056
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Graduation Interval
    Availability
    Price
  • 10 ml
    35 mm
    25 mm
    2 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 28,70 €
    28,70 €
    1 * 12 ST
       28,70 €
  • 25 ml
    47 mm
    31 mm
    5 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 28,70 €
    28,70 €
    1 * 12 ST
       28,70 €
  • 50 ml
    60 mm
    40 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 33,10 €
    33,10 €
    1 * 12 ST
       33,10 €
  • 100 ml
    70 mm
    49 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 34,40 €
    34,40 €
    1 * 12 ST
       34,40 €
  • 150 ml
    80 mm
    56 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 49,70 €
    49,70 €
    1 * 12 ST
       49,70 €
  • 250 ml
    94 mm
    68 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 24,90 €
    24,90 €
    1 * 6 ST
       24,90 €
  • 400 ml
    109 mm
    77 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 32,20 €
    32,20 €
    1 * 6 ST
       32,20 €
  • 500 ml
    122 mm
    88 mm
    50 ml
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 33,50 €
    33,50 €
    1 * 6 ST
       33,50 €
  • 600 ml
    125 mm
    91 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 34,10 €
    34,10 €
    1 * 6 ST
       34,10 €
  • 1000 ml
    149 mm
    102 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 47,10 €
    47,10 €
    1 * 6 ST
       47,10 €
  • 2000 ml
    183 mm
    133 mm
    200 ml
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 84,70 €
    84,70 €
    1 * 6 ST
       84,70 €
  • 3000 ml
    214 mm
    174 mm
    200 ml
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 109,00 €
    109,00 €
    1 * 4 ST
       109,00 €
  • 5000 ml
    248 mm
    185 mm
    500 ml
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 142,00 €
    142,00 €
    1 * 4 ST
       142,00 €