Orders LinkContactUsLinkComponent
Volumetric flasks, class B
Volumetric flasks, class B
 :   HIRSCHMANN
Volumetric flasks, class B
Volumetric flasks, class B
Volumetric flasks, class B
PMP, translucent, with PP NS-stopper.
Product Family Options
  • Capacity
    NS
    Height
  • 25 ml
    10/19
    115 mm
    1 * 2 ST
    1 * 2 ST - 29,10 €
    29,10 €
    1 * 2 ST
       29,10 €
  • 50 ml
    10/19
    150 mm
    1 * 2 ST
    1 * 2 ST - 34,50 €
    34,50 €
    1 * 2 ST
       34,50 €
  • 100 ml
    14/23
    180 mm
    1 * 2 ST
    1 * 2 ST - 39,00 €
    39,00 €
    1 * 2 ST
       39,00 €
  • 250 ml
    19/26
    235 mm
    1 * 2 ST
    1 * 2 ST - 48,60 €
    48,60 €
    1 * 2 ST
       48,60 €
  • 500 ml
    19/26
    270 mm
    1 * 2 ST
    1 * 2 ST - 50,30 €
    50,30 €
    1 * 2 ST
       50,30 €
  • 1000 ml
    24/29
    310 mm
    1 * 1 ST
    1 * 1 ST - 29,10 €
    29,10 €
    1 * 1 ST
       29,10 €
  • 10 ml
    10/19
    90 mm
    1 * 2 ST
    1 * 2 ST - 27,60 €
    27,60 €
    1 * 2 ST
       27,60 €