Measuring cylinders, tall form, class B, PMP
Supplier: HIRSCHMANN
Certificates
About this item
PMP, tall form, class B.
- Calibrated to contain (TC, In)
- With embossed scale (embossed scale recommended for autoclaving)
- Exposure to temperatures up to +121 °C will not cause permanent exceeding of tolerance limits
- Hexagonal socket, with spout
Certifications: DIN 12 681/ISO 6706
Frequently Bought Together