Condensers, Allihn, for Soxleth extractors
Supplier: Lenz Laborglas GmbH & CO.KG
Certificates
About this item
DURANĀ® borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With NS cone. Water connection with two threaded GL 14 plastic olives.
Frequently Bought Together