Order Entry
Belgium
ContactUsLinkComponent
Labelling and masking tapes, Post-it®
Labelling and masking tapes, Post-it®
Catalog # MMMA651H
Supplier:  3M
undefined
Labelling and masking tapes, Post-it®
Catalog # MMMA651H
Supplier:  3M

Specifications

  • Width:
    4,2 mm
  • Description:
    Labeling and masking tape, for 1 line
  • Environmentally Preferable:
  • Pk:
    1 Roll

Specifications

About this item

Repositionable acrylic. Post-it® Labelling and cover-up tape covers mistakes neatly. It is wide enough to cover one line of standard-sized type.

  • Easy to write on
  • Virtually invisible on copies

Length: 18,0 m.

Colour: White.