Recirculating coolers, SemiChill series
Fournisseur: JULABO GmbH
Certificats
À propos de cet article
Recirculating coolers can handle virtually any cooling requirements in laboratories or industrial environments efficiently.
- Environmentally friendly operation with low energy consumption
- Large, bright LED display with splash-proof keypad
- Optional air or water cooling with easy filling and drain tap easily accessible
- Circulating pumps with flow rates up to 60 L/min and 6 bar pressure
Frequently Bought Together