Orders LinkContactUsLinkComponent
Burette tubes without stopcocks
Burette tubes without stopcocks
Supplier:   HIRSCHMANN
Burette tubes without stopcocks
Supplier:  HIRSCHMANN
Certificates
Burette tubes without stopcocks
Burette tubes without stopcocks
3 Options Available Below
Product Details & Documents
Burette tube, class B, DURAN® borosilicate glass, with blue graduation, calibrated for EX.
  • Head at bottom
  • Stopcock attachment, 8 mm Ø
  • Suitable for replaceable stopcock
Product Family Options
Product Information
  • Capacity
    Division
    Tolerance
    Availability
    Price
  • 10 ml
    0,02 ml
    0,03 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,30 €
    25,30 €
    1 * 1 ST
       25,30 €
  • 25 ml
    0,02 ml
    0,045 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,30 €
    25,30 €
    1 * 1 ST
       25,30 €
  • 50 ml
    0,1 ml
    0,075 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,30 €
    25,30 €
    1 * 1 ST
       25,30 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • PTFE stopcock with PP tip
    PTFE stopcock with PP tip
    HIRS8901076
    Log in for availability
    1 * 1 ST - 28,60 €