Saisie de commande
Belgium
ContactUsLinkComponent
Pièce de distillation pour distillation directe
Pièce de distillation pour distillation directe
Catalog # LENZ05058001
undefined
Pièce de distillation pour distillation directe
Catalog # LENZ05058001
Specifications
  • Rodage mâle:
    14/23 NS
  • Rodage femelle:
    14/23 NS
  • Cdt:
    1
Specifications
About this item
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Short condensate path due to compact design of condenser and vacuum connection, with two cones NS 14/23, one socket NS 14/23 and three DIN threads GL 14.