Orders LinkContactUsLinkComponent
Multiple adapters
Multiple adapters
Supplier:   Lenz Laborglas
Multiple adapters
Supplier:  Lenz Laborglas
Certificates
Multiple adapters
Multiple adapters
2 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With one standard ground NS14/23 or NS 29/32 socket and two standard ground NS 14/23 or NS 29/32 cons. Side neck descending angle of 75°.
Documents
Product Family Options
Product Information
  • Cone size
    Socket size
    Availability
    Price
  • 14/23 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,30 €
    44,30 €
    1 * 1 ST
       44,30 €
  • 29/32 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,90 €
    42,90 €
    1 * 1 ST
       42,90 €