Flasks, round bottom with four necks and standard ground joints, side necks parallel
Fournisseur: Lenz Laborglas GmbH & CO.KG
Certificats
À propos de cet article
DURAN®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
Certifications: DIN 12392
Cork rings for round bottom flasks
Informations sur le produit
Frequently Bought Together