Orders LinkContactUsLinkComponent
Stopcocks for burettes, lateral
Stopcocks for burettes, lateral
Supplier:   Lenz Laborglas
Stopcocks for burettes, lateral
Supplier:  Lenz Laborglas
Certificates
Stopcocks for burettes, lateral
Stopcocks for burettes, lateral
2 Options Available Below
Product Details & Documents
NS stopcock made of borosilicate glass, facetted tip, with plug of solid glass or PTFE and screwthread retaining device.
  • Excellent chemical resistance
  • Excellent temperature resistance
Documents
Product Family Options
Product Information
  • Hole Ø
    Spigot
    Availability
    Price
  • 1,5 mm
    12,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,00 €
    53,00 €
    1 * 1 ST
       53,00 €
  • 1,5 mm
    12,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,70 €
    41,70 €
    1 * 1 ST
       41,70 €