Orders LinkContactUsLinkComponent
Stopcocks, two-way
Stopcocks, two-way
Supplier:   Lenz Laborglas
Stopcocks, two-way
Supplier:  Lenz Laborglas
Certificates
Stopcocks, two-way
Stopcocks, two-way
4 Options Available Below
Product Details & Documents
Stopcocks are made of DURAN® borosilicate glass with solid NS glass plug, screwthread retaining device, and two oblique bores.
  • Excellent chemical resistance
  • Excellent temperature resistance
With two oblique bores.
Leakage rate accoring to DIN 12540-1.
Documents
Product Family Options
With solid glass key
Product Information
  • Hole Ø
    Spigot
    Availability
    Price
  • 2,5 mm
    14,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 67,60 €
    67,60 €
    1 * 1 ST
       67,60 €
  • 4,0 mm
    18,8 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,70 €
    77,70 €
    1 * 1 ST
       77,70 €
With PTFE key
Product Information
  • Hole Ø
    Spigot
    Availability
    Price
  • 2,5 mm
    14,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 85,10 €
    85,10 €
    1 * 1 ST
       85,10 €
  • 4,0 mm
    18,8 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €