Order Entry
Belgium
ContactUsLinkComponent
2,2,4-Trimethylpentane, dehydrated (max. 0.01% H₂O) ≥99%, GPR RECTAPUR® for synthesis
Flammable
Health
Irritant
Environment
2,2,4-Trimethylpentane, dehydrated (max. 0.01% H₂O) ≥99%, GPR RECTAPUR® for synthesis
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Health
Irritant
Environment
2,2,4-Trimethylpentane, dehydrated (max. 0.01% H₂O) ≥99%, GPR RECTAPUR® for synthesis
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • IR Spectrum:
    Passes test
  • Density (20/4):
    0.690 - 0.695
  • Free acidity:
    Max. 0.0004 meq/g
  • Evaporation residue:
    Max. 50 ppm
  • Water:
    Max. 100 ppm

Specification Test Results

Frequently Bought Together