



2,2,4-Trimethylpentane 99% for spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:540-84-1
:
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943
:114.23 g/mol
Merck Index:13,05212
Boiling Pt:99.2 °C (1013 hPa)
Melting Pt:–107 °C
Density:0.69 g/cm³
Flash Pt:–12 °C
EINECS:208-759-1
UN:1262
ADR:3,II
:GHS07
:GHS08
:GHS09
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.02 %
- GC:≥98.5 %
- UV:at 210 nm A: ≤1
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
Specification Test Results
Frequently Bought Together