Orders LinkContactUsLinkComponent
Compact automatic burettes, SILBERBRAND
Compact automatic burettes, SILBERBRAND
Supplier:   Brand
Compact automatic burettes, SILBERBRAND
Supplier:  Brand
Certificates
Compact automatic burettes, SILBERBRAND
Compact automatic burettes, SILBERBRAND
2 Options Available Below
Product Details & Documents
Borosilicate glass 5.4, amber burette, amber PE bottle, class B.
  • Modular concept - all components are replaceable
  • Calibrated to deliver 'TD, Ex'
  • White graduations and automatic zeroing
25 ml: Error limit according to class B, DIN EN ISO 385.
50 ml: DIN EN ISO 385, class B.
Ordering information: Please order supports, burette clamps and rubber bellows separately.
Delivery information: Burette supplied with 1000 ml PE reservoir bottle with base and PVC filling tube.
Product Family Options
Product Information
  • Capacity
    Division
    Length
    Tolerance
    Availability
    Price
  • 25 ml
    0,1 ml
    495 mm
    0,08 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 176,00 €
    176,00 €
    1 * 1 ST
       176,00 €
  • 50 ml
    0,1 ml
    780 mm
    0,10 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 176,00 €
    176,00 €
    1 * 1 ST
       176,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Single bulb rubber bellows, orange coloured, with valve and tubing
    Single bulb rubber bellows, orange coloured, with valve and tubing
    BRND23400
    Log in for availability
    1 * 1 ST - 17,50 €
Product Information
  • Catalog #
    Capacity
    Availability
    Price
  • Bases for bottles
    Bases for bottles
    BRND23728
    1000 ml
    Log in for availability
    1 * 1 ST - 8,60 €
Product Information
  • Catalog #
    Capacity
    Availability
    Price
  • Bases for bottles
    Bases for bottles
    BRND23725
    500 ml
    Log in for availability
    1 * 1 ST - 7,20 €
Product Information
  • Catalog #
    Capacity
    Availability
    Price
  • Reservoir bottle, LDPE, brown, GL 28
    Reservoir bottle, LDPE, brown, GL 28
    BRND130260
    1000 ml
    Log in for availability
    1 * 1 ST - 9,40 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare filling tube for all compact automatic burettes, PVC
    Spare filling tube for all compact automatic burettes, PVC
    BRND11525
    Log in for availability
    1 * 1 ST - 4,80 €
Product Information
  • Catalog #
    Length
    Availability
    Price
  • Pump head, amber borosilicate glass 3.3, NS 29/32, fitting length 185 mm, nozzle outer Ø 7 mm,
    Pump head, amber borosilicate glass 3.3, NS 29/32, fitting length 185 mm, nozzle outer Ø 7 mm,
    BRND23876
    185 mm
    Log in for availability
    1 * 1 ST - 57,40 €
Product Information
  • Catalog #
    Capacity
    Division
    Availability
    Price
  • Spare burette tubes, amber glass
    Spare burette tubes, amber glass
    BRND23868
    50 ml
    0,1 ml
    Log in for availability
    1 * 1 ST - 72,00 €
Product Information
  • Catalog #
    Capacity
    Division
    Availability
    Price
  • Spare burette tubes, amber glass
    Spare burette tubes, amber glass
    BRND23866
    25 ml
    0,1 ml
    Log in for availability
    1 * 1 ST - 72,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare burette tip, amber borosilicate 5.4 glass, for 25 and 50 ml compact burette and compact automatic burette
    Spare burette tip, amber borosilicate 5.4 glass, for 25 and 50 ml compact burette and compact automatic burette
    BRND11515
    Log in for availability
    1 * 1 ST - 8,20 €