Orders LinkContactUsLinkComponent
Membrane filters, type 154
Membrane filters, type 154
   Sustainable Options Available
Supplier:   Sartorius
Membrane filters, type 154
Supplier:  Sartorius
   Sustainable Options Available
Certificates
Membrane filters, type 154
10 Options Available Below
Product Details & Documents
PES membrane filters are hydrophilic, have high flow rates thanks to the symmetric structure, have a low non-specific protein adsorption and are chemically resistant over a pH range of 1 to 14. They are recommended for the filtration of aqueous solutions as well for the protein filtration. Furthermore, the low level of extractables makes them suitable for environmental analysis.
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • 512-5076
  • SART15458-47-N
  • 512-2954
  • SART15407-142-MIG
  • SART15407-25-MIN
  • SART15407-47-MIN
  • SART15407-50-MIN
  • SART15406-25-N
  • SART15406-47-N
  • SART15406-50-N
  • Type:
  • 154
  • 154
  • 154
  • 154
  • 154
  • 154
  • 154
  • 154
  • 154
  • 154
  • Pore size:
  • 0,10 µm
  • 0,10 µm
  • 0,10 µm
  • 0,20 µm
  • 0,20 µm
  • 0,20 µm
  • 0,20 µm
  • 0,45 µm
  • 0,45 µm
  • 0,45 µm
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Thickness:
  • 150 μm
  • 150 μm
  • 150 μm
  • 150 μm
  • 150 μm
  • 150 μm
  • 150 μm
  • 150 μm
  • 150 μm
  • 150 μm
  • Water flow rate:
  • >7 ml/min/cm² (1 bar)
  • >7 ml/min/cm² (1 bar)
  • >7 ml/min/cm² (1 bar)
  • >28 ml/min/cm² (1 bar)
  • >28 ml/min/cm² (1 bar)
  • >28 ml/min/cm² (1 bar)
  • >28 ml/min/cm² (1 bar)
  • >32 ml/min/cm² (1 bar)
  • >32 ml/min/cm² (1 bar)
  • >32 ml/min/cm² (1 bar)
  • Environmental certification:
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
Specifications
Product Family Options
Product Information
  • Type
    Pore size
    Ø
    Availability
    Price
  • 154
    0,10 µm
    25 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 161,00 €
    161,00 €
    1 * 100 ST
       161,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    512-5076
    Type
    154
    Pore size
    0,10 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    150 μm
    Water flow rate
    >7 ml/min/cm² (1 bar)
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 154
    0,10 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 235,00 €
    235,00 €
    1 * 100 ST
       235,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SART15458-47-N
    Type
    154
    Pore size
    0,10 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    150 μm
    Water flow rate
    >7 ml/min/cm² (1 bar)
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 154
    0,10 µm
    50 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 245,00 €
    245,00 €
    1 * 100 ST
       245,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    512-2954
    Type
    154
    Pore size
    0,10 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    150 μm
    Water flow rate
    >7 ml/min/cm² (1 bar)
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 154
    0,20 µm
    142 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 252,00 €
    252,00 €
    1 * 25 ST
       252,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SART15407-142-MIG
    Type
    154
    Pore size
    0,20 µm
    Environmentally Preferable
    Thickness
    150 μm
    Water flow rate
    >28 ml/min/cm² (1 bar)
    Environmental certification
  • 154
    0,20 µm
    25 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 157,00 €
    157,00 €
    1 * 100 ST
       157,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SART15407-25-MIN
    Type
    154
    Pore size
    0,20 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    150 μm
    Water flow rate
    >28 ml/min/cm² (1 bar)
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 154
    0,20 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 235,00 €
    235,00 €
    1 * 100 ST
       235,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SART15407-47-MIN
    Type
    154
    Pore size
    0,20 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    150 μm
    Water flow rate
    >28 ml/min/cm² (1 bar)
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 154
    0,20 µm
    50 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 239,00 €
    239,00 €
    1 * 100 ST
       239,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SART15407-50-MIN
    Type
    154
    Pore size
    0,20 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    150 μm
    Water flow rate
    >28 ml/min/cm² (1 bar)
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 154
    0,45 µm
    25 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 157,00 €
    157,00 €
    1 * 100 ST
       157,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SART15406-25-N
    Type
    154
    Pore size
    0,45 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    150 μm
    Water flow rate
    >32 ml/min/cm² (1 bar)
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 154
    0,45 µm
    47 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 235,00 €
    235,00 €
    1 * 100 ST
       235,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SART15406-47-N
    Type
    154
    Pore size
    0,45 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    150 μm
    Water flow rate
    >32 ml/min/cm² (1 bar)
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 154
    0,45 µm
    50 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 245,00 €
    245,00 €
    1 * 100 ST
       245,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SART15406-50-N
    Type
    154
    Pore size
    0,45 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    150 μm
    Water flow rate
    >32 ml/min/cm² (1 bar)
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact