Order Entry
Belgium
Orders LinkContactUsLinkComponent
4-Hydroxy-4-methyl-2-pentanone ≥98%, TECHNICAL

4-Hydroxy-4-methyl-2-pentanone ≥98%, TECHNICAL

Glass bottle, 1 L
Catalog # 23424.294
Flammable
Irritant
Supplier:   VWR Chemicals
4-Hydroxy-4-methyl-2-pentanone ≥98%, TECHNICAL
Catalog # 23424.294
Supplier:  VWR Chemicals
CAS Number:  123-42-2
4-Hydroxy-4-methyl-2-pentanone ≥98%, TECHNICAL
4-Hydroxy-4-methyl-2-pentanone ≥98%, TECHNICAL

Glass bottle, 1 L

CAS Number:  123-42-2
Price
47,50 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Diacetone alcohol
  • CAS Number:123-42-2
  • Formula:(CH₃)₂C(OH)CH₂COCH₃
  • MDL Number:MFCD00004471
  • Molecular Weight:116.16 g/mol
  • Merck Index:13,02983
  • Boiling Pt:166 °C (1013 hPa)
  • Melting Pt:–44 °C
  • Density:0.94 g/cm³ (20 °C)
  • Flash Pt:58 °C
  • Storage Temperature:Ambient
  • EINECS:204-626-7
  • UN:1148
  • ADR:3,III
  • REACH:01-2119473975-21
Specifications

Specification Test Results

  • Assay:
    Min. 98 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.