4-Hydroxy-4-methyl-2-pentanone 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-42-2
:
Formula:(CH₃)₂C(OH)CH₂COCH₃
MDL Number:MFCD00004471
:116.16 g/mol
Merck Index:13,02983
Boiling Pt:166 °C (760 mmHg)
Melting Pt:–42.8 °C
Density:0.9385 g/cm³
Flash Pt:58 °C
Storage Temperature:Ambient
EINECS:204-626-7
UN:1148
ADR:3,III
REACH:01-2119473975-21
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4220 to 1.4250 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤10 APHA
Specification Test Results
Frequently Bought Together