Order Entry
Belgium
ContactUsLinkComponent
5-Methyl-3-heptanone for synthesis, Sigma-Aldrich®
Flammable
Irritant
5-Methyl-3-heptanone for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Irritant
5-Methyl-3-heptanone for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    3 g/l (20 °C)
  • Melting Point:
    -57 °C
  • Molar Mass:
    128.22 g/mol
  • Boiling Point:
    157 - 162 °C (1013 hPa)
  • Vapor Pressure:
    2.7 hPa (25 °C)
  • Flash Point:
    43 °C
  • Refractive Index:
    1.4209 (20 °C, 589 nm)
  • Explosion Limit:
    0.9 - 48 %(V)
  • Density:
    0.82 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together