Order Entry
Belgium
ContactUsLinkComponent
4-Methyl-2-pentanone, Supelco®
Flammable
Irritant
4-Methyl-2-pentanone, Supelco®
Catalog # 02474-5ML
Supplier:  Merck
CAS Number:  
undefined
Flammable
Irritant
4-Methyl-2-pentanone, Supelco®
Catalog # 02474-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :100.16 g/mol
Merck Index:13,05227
Boiling Pt:117.4 °C (1013 hPa)
Melting Pt:–84 °C
Density:0.801 g/cm³ (20 °C)
Flash Pt:16 °C
EINECS:203-550-1
UN:1245
ADR:3,II
 :M-Clarity™ quality level = MQ100
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:108-10-1
Formula:(CH₃)₂CHCH₂COCH₃
MDL Number:MFCD00008938