4-Methyl-2-pentanone 99+% for HPLC
:108-10-1
:
:(CH₃)₂CHCH₂COCH₃
:MFCD00008938
:100.16 g/mol
:14,5207
:117…118 °C
:–84 °C
:0.801
:13 °C (55 °F)
:Ambient
:203-550-1
:1245
:3,II
:GHS02
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Frequently Bought Together