all-rac-α-Tocopherol 96.0-102.0%, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, E307, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:10191-41-0
:1406-66-2
Formula:C₂₉H₅₀O₂
MDL Number:MFCD00072051
:430.71 g/mol
EINECS:233-466-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (GC, Ph. Eur.):96.0 - 102.0 %
- Assay (GC, USP):96.0 - 102.0 %
- Identity (IR):conforms
- Identity (GC):conforms
- Identity (colour reaction):conforms
- Appearance:clear, colourless to yellow-brownish, viscous, oily liquid; almost odourless
- Acidity:conforms
- Optical rotation α 20/D (100 g/l; anhydrous ethanol):-0.01 - 0.01 °
- Optical rotation α 25/D (100 g/l; chloroforme):-0.05 - 0.05 °
- Refractive index (n 20/D):1.503 - 1.507
- Spec. Absorptivity A 1%/1cm (292 nm; 0,1 g/l, anhydrous ethanol):72 - 76
- Spec. Absorptivity A1%/1cm (292 nm; 0,05 g/l , anhydrous Ethanol):71 - 76
- Heavy metals (as Pb):≤ 10 ppm
- As (Arsenic):≤ 3 ppm
- Cu (Copper):≤ 10 ppm
- Hg (Mercury):≤ 1 ppm
- Pb (Lead):≤ 2 ppm
- Zn (Zinc)#:≤ 25 ppm
- Heptane (HS-GC):≤ 5000 ppm
- Methanol (HS-GC):≤ 3000 ppm
- Pyridine (HS-GC):≤ 200 ppm
- Toluene (HS-GC):≤ 890 ppm
- Other residual solvents (ICH Q3C):excluded by manucfacturing process
- Related substances (Ph. Eur., impurity A):≤ 0.5 %
- Related substances (Ph. Eur., impurity B):≤ 1.5 %
- Related substances (Ph. Eur., sum of impurities C and D):≤ 1.0 %
- Related substances (Ph. Eur., any other impurity):≤ 0.25 %
- Related substances (Ph. Eur., total impurities):≤ 2.5 %
- Sulfated ash (600 °C):≤ 0.1 %
Specification Test Results
Frequently Bought Together