Order Entry
Belgium
Orders LinkContactUsLinkComponent
DL-α-Tocopherol (Vitamin E)

DL-α-Tocopherol (Vitamin E)

Glass bottle for solids, 1,5 mL
Catalog # EDQMT1550000
Supplier:   EDQM
DL-α-Tocopherol (Vitamin E)
Catalog # EDQMT1550000
Supplier:  EDQM
CAS Number:  10191-41-0
DL-α-Tocopherol (Vitamin E)

Glass bottle for solids, 1,5 mL

CAS Number:  10191-41-0
Price
270,00 €
Log in for availability
1 * 1,5 mL
Product Details & Documents
Synonyms:  (±)-α-Tocopherol, Vitamin E Alcohol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol, DL-alpha-Tocopherol, Vitamin E
  • CAS Number:10191-41-0
  • Formula:C₂₉H₅₀O₂
  • MDL Number:MFCD00072051
  • Molecular Weight:430.71 g/mol
  • EINECS:233-466-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1,5 mL

Specifications