Order Entry
Belgium
ContactUsLinkComponent
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
Supplier:  VWR Chemicals
Certificates
undefined
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 95,0%
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 0,02 %
  • Pesticide analysis (Lindane/ECD):
    Max. 5 ng/l

Specification Test Results

Frequently Bought Together