Order Entry
Belgium
ContactUsLinkComponent
n-Hexane ≥95%, TECHNICAL
Flammable
Health
Toxic
Environment
n-Hexane ≥95%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Health
Toxic
Environment
n-Hexane ≥95%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Assay:
    Min. 95 %
  • Boiling point:
    68 - 70 °C
  • Benzene:
    Max. 100 ppm
  • Evaporation residue:
    Max. 20 ppm

Specification Test Results

Frequently Bought Together