Order Entry
Belgium
ContactUsLinkComponent
n-Hexane ≥99%, AnalaR NORMAPUR® analytical reagent
Flammable
Health
Toxic
Environment
n-Hexane ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Health
Toxic
Environment
n-Hexane ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Acidity:
    Max. 0.0004 meq/g
  • Colouration:
    Max. 10 APHA
  • Density (20/4):
    0.658 - 0.662
  • Benzene:
    Max. 100 ppm
  • Evaporation residue:
    Max. 10 ppm
  • Thiophene:
    Max. 1 ppm
  • Total S (as SO4):
    Max. 50 ppm
  • Water:
    Max. 100 ppm
  • Al (Aluminium):
    Max. 0.5 ppm
  • B (Boron):
    Max. 0.02 ppm
  • Ba (Barium):
    Max. 0.1 ppm
  • Ca (Calcium):
    Max. 0.5 ppm
  • Cd (Cadmium):
    Max. 0.05 ppm
  • Co (Cobalt):
    Max. 0.02 ppm
  • Cr (Chromium):
    Max. 0.02 ppm
  • Cu (Copper):
    Max. 0.02 ppm
  • Fe (Iron):
    Max. 0.1 ppm
  • Mg (Magnesium):
    Max. 0.1 ppm
  • Mn (Manganese):
    Max. 0.02 ppm
  • Ni (Nickel):
    Max. 0.02 ppm
  • Pb (Lead):
    Max. 0.1 ppm
  • Sn (Tin):
    Max. 0.1 ppm
  • Zn (Zinc):
    Max. 0.1 ppm

Specification Test Results

Frequently Bought Together