Order Entry
Belgium
ContactUsLinkComponent
n-Hexane, dehydrated (max. 0.01% H₂O) ≥95%, GPR RECTAPUR® for synthesis
n-Hexane, dehydrated (max. 0.01% H₂O) ≥95%, GPR RECTAPUR® for synthesis
Supplier:  VWR Chemicals
Certificates
undefined
n-Hexane, dehydrated (max. 0.01% H₂O) ≥95%, GPR RECTAPUR® for synthesis
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 95 %
  • Evaporation residue:
    Max. 50 ppm
  • Water:
    Max. 100 ppm

Specification Test Results

Frequently Bought Together