



n-Hexane ≥95%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
:110-54-3
:92112-69-1
:H₃C(CH₂)₄CH₃
:MFCD00009520
:86.18 g/mol
:13,04712
:69 °C (1013 hPa)
:–94,3 °C
:0.659 g/cm³ (20 °C)
:–22 °C
:203-777-6
:1208
:3,II
:01-2119480412-44
:GHS02
:GHS08
:GHS06
:GHS09
:Ambient
- Assay (n-Hexane):Min. 95.0 %
- Assay (Isomers):Min. 98.5 %
- IR Spectrum:Passes test
- Thiophene:Passes test
- Acidity:Max. 0.0002 meq/g
- Boiling point:68 - 69 °C
- Bromine value:Max. 0.5
- Colouration:Max. 10 APHA
- Density (20/4):0.658 - 0.662
- Density (20/20):0.659 - 0.663
- Refractive index (20°C):1.375 - 1.376
- Substances coloured by H2SO4:Max. 90 APHA
- Benzene:Max. 100 ppm
- Evaporation residue:Max. 10 ppm
- Total S (as SO4):Max. 10 ppm
- Water:Max. 100 ppm
- Al (Aluminium):Max. 0.5 ppm
- B (Boron):Max. 0.02 ppm
- Ba (Barium):Max. 0.1 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.01 ppm
- Co (Cobalt):Max. 0.02 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.01 ppm
- Fe (Iron):Max. 0.1 ppm
- K (Potassium):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.1 ppm
- Mn (Manganese):Max. 0.02 ppm
- Na (Sodium):Max. 0.1 ppm
- Ni (Nickel):Max. 0.01 ppm
- Pb (Lead):Max. 0.01 ppm
- Sn (Tin):Max. 0.1 ppm
- Sr (Strontium):Max. 0.02 ppm
- Zn (Zinc):Max. 0.01 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Aluminium bottle for liquids DIN 45 UN X (ZPACK8000430)
Frequently Bought Together