



n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94.3 °C
Density:0,659 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:–22 °C (closed cup)
:GHS02
:GHS08
:GHS07
:GHS09
Related Information
Frequently Bought Together