Order Entry
Belgium
ContactUsLinkComponent
 
n-Hexane ≥95% (by GC), CHROMASOLV™ for pesticide residue analysis, for analysis of dioxins, furans and PCB, Riedel-de Haën™
 
undefined
n-Hexane ≥95% (by GC), CHROMASOLV™ for pesticide residue analysis, for analysis of dioxins, furans and PCB, Riedel-de Haën™

 

  • Assay (GC):
    Min. 95 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Suitability for GC-MS of dioxines, furanes, PCPs:
    complying
  • Vapor Density:
    ~3 (vs air)
  • Vapor Pressure:
    ~132 mmHg ( 20 °C)
  • Refractive Index:
    n20/D 1.375 (lit.)
  • Explosion Limit:
    7.7 %

 

 

Frequently Bought Together