Order Entry
Belgium
ContactUsLinkComponent
Hexane (mixture of isomers) ≥99.5% (by GC, corrected for water content) (n-hexane ≥95%), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Flammable
Health
Irritant
Environment
Hexane (mixture of isomers) ≥99.5% (by GC, corrected for water content) (n-hexane ≥95%), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Flammable
Health
Irritant
Environment
Hexane (mixture of isomers) ≥99.5% (by GC, corrected for water content) (n-hexane ≥95%), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 5
  • FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):
    ≤ 5
  • ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):
    ≤ 10
  • ECD-Sensitive Impurities (as Ethylene Dibromide) - Single Impurity Peak (ng/mL):
    ≤ 5
  • Assay (Total Saturated C₆ Isomers) (by GC, corrected for water):
    ≥ 99.5 %
  • Assay (as n-Hexane) (by GC, corrected for water):
    ≥ 95 %
  • Color (APHA):
    ≤ 10
  • Residue after Evaporation:
    ≤ 1.0 ppm
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Water (by KF, coulometric):
    ≤ 0.05 %

Specification Test Results

Frequently Bought Together