
n-Hexadecane
Catalog # EHERC14191500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:226.45 g/mol
Boiling Pt:287 °C (1013 hPa)
Melting Pt:18 °C
Density:0.77331 g/cm³ (20 °C)
Flash Pt:135 °C (closed cup)
Storage Temperature:Ambient
EINECS:208-878-9
:GHS08
CAS Number:544-76-3
Formula:H₃C(CH₂)₁₄CH₃
MDL Number:MFCD00008998