

n-Hexadecane ≥99.5%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:544-76-3
:
Formula:H₃C(CH₂)₁₄CH₃
MDL Number:MFCD00008998
:226.45 g/mol
Boiling Pt:287 °C
Melting Pt:18 °C
Density:0.773
Flash Pt:135 °C (275 °F)
Storage Temperature:Ambient
EINECS:208-878-9
:GHS08
:GHS07
Related Information
Frequently Bought Together